Order Entry
Export
Orders LinkContactUsLinkComponent
 

WHEATON® Sample Vials in Lab File®

Shorty vials in Lab File™ without caps, 2,00 ml, 15×28 mm, Clear, No, 13-425
  218-2215
WHEATON® Sample Vials in Lab File®
  218-2215
WHEATON® Sample Vials in Lab File®
WHEATON® Sample Vials in Lab File®

Shorty vials in Lab File™ without caps, 2,00 ml, 15×28 mm, Clear, No, 13-425

 
209,00 €
   
1 * 200 items
 

Clear vials made from low extractable borosilicate glass, amber vials made from borosilicate glass.

  • Sample vials with caps attached to vial help maintain cleanliness
  • Lab File® with partitioned trays provides an easy way to inventory samples or to store empty vials
  • Clear vials conform to ASTM E438 Type I, Class A and USP Type I requirements
  • Amber vials conform to USP Type I requirements for light transmission to protect light-sensitive products
  • 14B rubber lined black phenolic screw cap for aqueous samples
 
  • Cap included:
    No
  • Cap Size:
    13-425
  • Ø×H:
    15×28 mm
  • Capacity:
    2,00 ml
  • Colour:
    Clear
  • Description:
    Shorty vials in Lab File™ without caps
  • Seq:
    18