Order Entry
Export
Orders LinkContactUsLinkComponent
 

Stoppers, LDPE

Natural, 17,5 mm, 19 mm
  217-9251
 :   SEMADENI
Stoppers, LDPE
  217-9251
 :  SEMADENI
Stoppers, LDPE
Stoppers, LDPE

Natural, 17,5 mm, 19 mm

 
35,10 €
   
1 * 100 items
 

Lamellar stoppers made from soft PE-LD.

  • Practical grip edge
  • Bright tolerance through soft lamellars

 

 
  • Ø bottom:
    17,5 mm
  • Ø top:
    19 mm
  • Colour:
    Natural
  • Seq:
    11