Order Entry
Export
Orders LinkContactUsLinkComponent
 

Stoppers, LDPE

Natural, 12,5 mm, 14 mm
Catalog # 217-9247
Supplier:   SEMADENI
Stoppers, LDPE
Catalog # 217-9247
Supplier:  SEMADENI
Stoppers, LDPE
Stoppers, LDPE

Natural, 12,5 mm, 14 mm

Price
13,00 €
Log in for availability
1 * 100 items
Product Details & Documents

Lamellar stoppers made from soft PE-LD.

  • Practical grip edge
  • Bright tolerance through soft lamellars

 

Specifications
  • Ø bottom:
    12,5 mm
  • Ø top:
    14 mm
  • Colour:
    Natural
  • Seq:
    06