Instrument trays
370×240×80 mm
Catalog # 216-7401
Supplier: DELTALAB
Jump to:
Product Details & Documents
Made of stainless steel.
- Excellent chemical resistance
- Excellent temperature resistance
- Easy to clean
Specifications
- Seq:01
- W×D×H:370×240×80 mm