Orders LinkContactUsLinkComponent
Wire basket, round
Wire basket, round
230×180 mm
Catalog # 216-7257
Supplier:   DELTALAB
Wire basket, round
Catalog # 216-7257
Supplier:  DELTALAB
Wire basket, round
Wire basket, round
230×180 mm
Price
256,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel.
These round baskets feature a folding handle for more convenience. The base of these baskets is reinforced by a thick mesh for added strength.
Ordering information: Models with inner crossed dividers are available.
Documents
Specifications
  • Ø×H:
    230×180 mm
  • Seq:
    03