Bottles, serum, WHEATON®
3,00 ml, 17×38 mm, Natural, No, 13 mm
216-3019
These bottles are manufactured from 400 borosilicate glass that conforms to USP Type I requirements.
- Accepts standard stoppers and aluminum seals
- PP bottle is used extensively for diluent solutions
- HDPE bottle is widely used for animal health products and storage of non-medical products
- Autoclavable at 121 °C for 20 minutes
- Cap included:No
- Cap Size:13 mm
- Ø×H:17×38 mm
- Capacity:3,00 ml
- Colour:Natural
- Seq:03



