Orders LinkContactUsLinkComponent
Instrument dishes
Instrument dishes
Lid for dishes 216-1403, 200x100x10 mm
Catalog # 216-1413
Instrument dishes
Catalog # 216-1413
Instrument dishes
Instrument dishes
Lid for dishes 216-1403, 200x100x10 mm
Price
19,70 €
Log in for availability
1 * 1 items
Product Details & Documents
Thick-walled glass.
  • Can be sterilised in hot air up to 180 °C
  • Robust
  • Stainless steel lids are available separately
Multipurpose dishes with vertical walls for a variety of applications in the laboratory, field use and industry.
Specifications
  • Description:
    Lid for dishes 216-1403
  • Seq:
    10
  • W×D×H:
    200x100x10 mm