Orders LinkContactUsLinkComponent
Watch glasses
Watch glasses
Watch glass, soda glass, with fused edges, 100 mm
Catalog # 216-1178
Supplier:   KAVALIER GLASS
Watch glasses
Catalog # 216-1178
Supplier:  KAVALIER GLASS
Watch glasses
Watch glasses
Watch glass, soda glass, with fused edges, 100 mm
Price
5,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Soda-lime glass.
  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses
  • Quick and easy to clean
With fused edges.
Certifications: DIN 12341
Documents
Specifications
  • Ø:
    100 mm
  • Description:
    Watch glass, soda glass, with fused edges
  • Seq:
    13