Orders LinkContactUsLinkComponent
Bottles, square, narrow neck, rectangular, HDPE, with thread, 310 series
1000 ml, 25 mm, 39,2 mm, 84×66×222,5 mm
  215-3304
 :   Kautex
Bottles, square, narrow neck, rectangular, HDPE, with thread, 310 series
  215-3304
 :  Kautex
Bottles, square, narrow neck, rectangular, HDPE, with thread, 310 series
Bottles, square, narrow neck, rectangular, HDPE, with thread, 310 series
1000 ml, 25 mm, 39,2 mm, 84×66×222,5 mm
2,80 €
1 * 1 items
HDPE, rectangular, natural colour.
  • High stability
  • Recessed labelling field
  • Leakproof
  • Suitable for food
  • Halal certified
Ribbed sides for secure grip. Four different types of closures.
  • Thread:
    25 mm
  • Capacity:
    1000 ml
  • W×D×H:
    84×66×222,5 mm
  • Neck I-Ø:
    39,2 mm