Bottles, square, narrow neck, rectangular, HDPE, with thread, 310 series
500 ml, 25 mm, 19,0 mm, 63,5×54,5×180 mm
Catalog # 215-3303
Supplier: Kautex
Bottles, square, narrow neck, rectangular, HDPE, with thread, 310 series
Catalog # 215-3303
Supplier: Kautex
Jump to:
Product Details & Documents
HDPE, rectangular, natural colour.
- High stability
- Recessed labelling field
- Leakproof
- Suitable for food
- Halal certified
Ribbed sides for secure grip. Four different types of closures.
Specifications
- Thread:25 mm
- Capacity:500 ml
- W×D×H:63,5×54,5×180 mm
- Neck I-Ø:19,0 mm



