Bottles, square, narrow neck, rectangular, HDPE, with thread, 310 series
250 ml, 25 mm, 19,0 mm, 53,5×42,5×148 mm
Catalog # 215-3302
Supplier: Kautex
Bottles, square, narrow neck, rectangular, HDPE, with thread, 310 series
Catalog # 215-3302
Supplier: Kautex
Jump to:
Product Details & Documents
HDPE, rectangular, natural colour.
- High stability
- Recessed labelling field
- Leakproof
- Suitable for food
- Halal certified
Ribbed sides for secure grip. Four different types of closures.
Specifications
- Thread:25 mm
- Capacity:250 ml
- W×D×H:53,5×42,5×148 mm
- Neck I-Ø:19,0 mm



