Orders LinkContactUsLinkComponent
Bottles, wide neck, square
Bottles, wide neck, square
500 ml, GL 54
Catalog # 215-1676
Supplier:   witeg Labortechnik
Bottles, wide neck, square
Catalog # 215-1676
Supplier:  witeg Labortechnik
Bottles, wide neck, square
Bottles, wide neck, square
500 ml, GL 54
Price
13,50 €
Log in for availability
1 * 1 items
Product Details & Documents
Soda-lime glass, coated.
  • With PU coating to protect the bottle from scratches, splinters, and leaking liquids
Without cap and pouring ring.
Specifications
  • Thread:
    GL 54
  • Capacity:
    500 ml