Push-in caps for tubes
LDPE, Caps, Ø 46 mm, Natural
Catalog # 215-0217
Jump to:
Discontinued
Product Details & Documents
LDPE, translucent.
Caps with rim for sealing test tubes, flasks and bottles.
Specifications
- Colour:Natural
- Description:Caps, Ø 46 mm
- Material:LDPE
- Seq:22