Benzalkonium chloride
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 63449-41-2 Molecular Formula: C6H5CH2N(CH3)2RCl Physical Form: Semi-solid/solid, R=C8H17 to C18H37 Solubility: Soluble in water Merck Reference: 14,1059 UN No.: 2923 MDL No.: MFCD00145757
Cationic surface active agent
Synonyms: Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride
- CAS Number:63449-41-2
- Formula:C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
- MDL Number:MFCD00145757
- Merck Index:14,1059
- Melting Pt:29…34 °C
- Density:0.98 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:264-151-6
- UN:2923
- ADR:8,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



