Flasks, round bottom, with standard ground joint
500 ml, 29/32 NS, 177 mm, 107 mm
214-1423
: VITLAB
PFA, translucent.
- Very good chemical resistance
- Excellent thermal resistance from −270 to +250 °C
- Autoclavable
Can also be used as safety flasks for rotary evaporators.
- Ext.Ø:107 mm
- Socket size:29/32 NS
- Capacity:500 ml
- Height:177 mm



