8-Quinolinol ACS
Glass bottle for solids, 50 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 148-24-3 Molecular Formula: C9H7NO Formula Weight: 145.16 Physical Form: Granular Sensitivity: Light Sensitive Appearance: White to pale yellow Melting Point: 72-76° Boiling Point: 267° Solubility: Almost insoluble in water and diethyl ether. Highly soluble in ethanol, acetone, chloroform, and benzene. Beilstein Reference: 114512 Merck Reference: 14,4843 MDL No.: MFCD00006807
A bacteriostatic and fungistatic agent. Also a chelating agent for metals
Synonyms: 8-Hydroxyquinoline
- CAS Number:148-24-3
- Formula:C₉H₇NO
- MDL Number:MFCD00006807
- Molecular Weight:145.16 g/mol
- Merck Index:14,4843
- Boiling Pt:267 °C
- Melting Pt:72…76 °C
- Density:1.03 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:205-711-1
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



