Potassium bromide for spectroscopy
1 lb
Supplier: Thermo Fisher Scientific
Potassium bromide for spectroscopy
Catalog # 39796.P2
Supplier: Thermo Fisher Scientific
CAS Number: 7758-02-3
Jump to:
Product Details & Documents
CAS No.: 7758-02-3 Molecular Formula: KBr Formula Weight: 119.01 Physical Form: Random Cuttings, Chunks Density: 2.75 Refractive Index: 1.559 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 730° Boiling Point: 1435° Merck Reference: 14,7618 MDL No.: MFCD00011358
- CAS Number:7758-02-3
- Formula:KBr
- MDL Number:MFCD00011358
- Molecular Weight:119 g/mol
- Merck Index:14,7618
- Boiling Pt:1435 °C
- Melting Pt:730 °C
- Density:2.75
- Storage Temperature:Ambient
- EINECS:231-830-3
Specifications
Specifications
- Pk:1 lb



