Order Entry
Export
Orders LinkContactUsLinkComponent
Caffeine ≥99.7%

Caffeine ≥99.7%

Glass bottle for solids, 500 g
Catalog # 39214.36
Irritant
Caffeine ≥99.7%
Catalog # 39214.36
CAS Number:  58-08-2
Caffeine ≥99.7%
Caffeine ≥99.7%

Glass bottle for solids, 500 g

CAS Number:  58-08-2
Price
306,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99.7% CAS No.: 58-08-2 Molecular Formula: C8H10N4O2 Formula Weight: 194.19 Physical Form: Powder Density: 1.230 Appearance: White Melting Point: 234-236° Boiling Point: 178° subl. Beilstein Reference: 17705 Merck Reference: 14,1636 UN No.: 1544 MDL No.: MFCD00005758

Synonyms:  3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, Methyltheobromine, 1,3,7-Trimethyl-2,6-dioxopurine, 2,6-Dihydroxy-1,3,7-trimethylpurine
  • CAS Number:58-08-2
  • Formula:C₈H₁₀N₄O₂
  • MDL Number:MFCD00005758
  • Molecular Weight:194.19 g/mol
  • Merck Index:14,1636
  • Boiling Pt:178 °C (subl.)
  • Melting Pt:234…236 °C
  • Density:1.230
  • Storage Temperature:Ambient
  • EINECS:200-362-1
  • UN:1544
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications