Caffeine ≥99.7%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99.7% CAS No.: 58-08-2 Molecular Formula: C8H10N4O2 Formula Weight: 194.19 Physical Form: Powder Density: 1.230 Appearance: White Melting Point: 234-236° Boiling Point: 178° subl. Beilstein Reference: 17705 Merck Reference: 14,1636 UN No.: 1544 MDL No.: MFCD00005758
Synonyms: 3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, Methyltheobromine, 1,3,7-Trimethyl-2,6-dioxopurine, 2,6-Dihydroxy-1,3,7-trimethylpurine
- CAS Number:58-08-2
- Formula:C₈H₁₀N₄O₂
- MDL Number:MFCD00005758
- Molecular Weight:194.19 g/mol
- Merck Index:14,1636
- Boiling Pt:178 °C (subl.)
- Melting Pt:234…236 °C
- Density:1.230
- Storage Temperature:Ambient
- EINECS:200-362-1
- UN:1544
- ADR:6.1,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



