n-Hexane ≥95% for HPLC
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 95% min CAS No.: 110-54-3 Molecular Formula: CH3(CH2)4CH3 Formula Weight: 86.18 Flash Point: -23°(-9°F) Physical Form: Liquid, Distilled in Glass Density: 0.659 Refractive Index: 1.3750 Odour: Weak, characteristic Melting Point: -95° Boiling Point: 69° Beilstein Reference: 1730733 Merck Reference: 14,4694 UN No.: 1208 MDL No.: MFCD00009520
Synonyms: Hexyl hydride, Hexanen, Hexane
- CAS Number:110-54-3
- Formula:H₃C(CH₂)₄CH₃
- MDL Number:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:14,4694
- Boiling Pt:69 °C
- Melting Pt:–95 °C
- Density:0.659
- Flash Pt:–23 °C (–9 °F)
- Storage Temperature:Ambient
- EINECS:203-777-6
- UN:1208
- ADR:3,II
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L



