1,4-Dioxane 99+% ACS
Glass bottle, 4 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99+% CAS No.: 123-91-1 Molecular Formula: C4H8O2 Formula Weight: 88.11 Flash Point: 12°(54°F) Physical Form: Liquid Density: 1.034 Refractive Index: 1.4220 Sensitivity: Hygroscopic Odour: Ether-like Melting Point: 11.8° Boiling Point: 100-102° Beilstein Reference: 102551 Merck Reference: 14,3300 UN No.: 1165 MDL No.: MFCD00006571
Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide
- CAS Number:123-91-1
- Formula:C₄H₈O₂
- MDL Number:MFCD00006571
- Molecular Weight:88.11 g/mol
- Merck Index:14,3300
- Boiling Pt:100…102 °C
- Melting Pt:11.8 °C
- Density:1.034
- Flash Pt:12 °C (54 °F)
- Storage Temperature:Ambient
- EINECS:204-661-8
- UN:1165
- ADR:3,II
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:4 L



