Phenolphthalein ACS
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 77-09-8 Molecular Formula: C20H14O4 Formula Weight: 318.33 Physical Form: Powder Density: 1.299 Melting Point: 258-262° Beilstein Reference: 284423 Merck Reference: 14,7243 MDL No.: MFCD00005913
Indicator
Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
- CAS Number:77-09-8
- Formula:C₂₀H₁₄O₄
- MDL Number:MFCD00005913
- Molecular Weight:318.33 g/mol
- Merck Index:14,7243
- Melting Pt:258…262 °C
- Density:1.299
- Storage Temperature:Ambient
- EINECS:201-004-7
- REACH:01-2119498295-24
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



