Infrared optical windows
Crystal optic rectangle, polished both sides, Potassium bromide, 30 × 15 mm, 4 mm
These optical crystal windows are ideal for infrared spectroscopy.
- Disc or rectangular
- Polished or unpolished
A polishing kit is available for those who wish to polish their windows.
- Window material:Potassium bromide
- Description:Crystal optic rectangle, polished both sides
- Dimensions:30 × 15 mm
- Seq:65
- Thickness:4 mm



