Infrared optical windows
Crystal optic disc, polished both sides, Sodium chloride, 13 mm, 1 mm
These optical crystal windows are ideal for infrared spectroscopy.
- Disc or rectangular
- Polished or unpolished
A polishing kit is available for those who wish to polish their windows.
- Window material:Sodium chloride
- Description:Crystal optic disc, polished both sides
- Dimensions:13 mm
- Seq:89
- Thickness:1 mm



