Infrared optical windows
Crystal optic disc, polished both sides, Barium fluoride, 25 mm, 4 mm
These optical crystal windows are ideal for infrared spectroscopy.
- Disc or rectangular
- Polished or unpolished
A polishing kit is available for those who wish to polish their windows.
- Window material:Barium fluoride
- Description:Crystal optic disc, polished both sides
- Dimensions:25 mm
- Seq:41
- Thickness:4 mm



