Order Entry
Export
Orders LinkContactUsLinkComponent
Lead(II)-2-ethylhexanoate

Lead(II)-2-ethylhexanoate

250 g
Catalog # 36711.30
Health
Irritant
Lead(II)-2-ethylhexanoate
Catalog # 36711.30
CAS Number:  301-08-6
Lead(II)-2-ethylhexanoate
Lead(II)-2-ethylhexanoate

250 g

CAS Number:  301-08-6
Price
135,00 €
Log in for availability
1 * 250 g
Product Details & Documents

CAS No.: 301-08-6 Molecular Formula: C16H30O4Pb Formula Weight: 493.61 Physical Form: Viscous Liquid Density: 1.56 Odour: Odourless UN No.: UN3082 MDL No.: MFCD00014003

Synonyms:  Lead(II) 2-ethylhexanoate
  • CAS Number:301-08-6
  • Formula:C₁₆H₃₀O₄Pb
  • MDL Number:MFCD00014003
  • Molecular Weight:493.61 g/mol
  • Density:1.56
  • Storage Temperature:Ambient
  • EINECS:206-107-0
  • ADR:9,III
Specifications

Specifications

  • Pk:
    250 g

Specifications