Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,3-Dichloropropane ≥95%

Glass bottle for solids, 25 g
Catalog # 36685.14
Flammable
1,3-Dichloropropane ≥95%
Catalog # 36685.14
CAS Number:  142-28-9
1,3-Dichloropropane ≥95%
1,3-Dichloropropane ≥95%

Glass bottle for solids, 25 g

CAS Number:  142-28-9
Price
102,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Trimethylendichloride
  • CAS Number:142-28-9
  • Formula:ClH₂CCH₂CH₂Cl
  • MDL Number:MFCD00000999
  • Molecular Weight:112.99 g/mol
  • Boiling Pt:120 °C
  • Melting Pt:–99.5 °C
  • Density:1.190
  • Flash Pt:20 °C (68 °F)
  • Storage Temperature:Ambient
  • EINECS:205-531-3
  • ADR:3,II
  •  :Density: 1.1717 g/cm³ (20 °C)
  • UN:1993
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications