Order Entry
Export
Orders LinkContactUsLinkComponent
Acrylamide ≥99.9%

Acrylamide ≥99.9%

Glass bottle for solids, 100 g
Catalog # 36545.22
Toxic
Health
Irritant
Acrylamide ≥99.9%
Catalog # 36545.22
CAS Number:  79-06-1
Acrylamide ≥99.9%
Acrylamide ≥99.9%

Glass bottle for solids, 100 g

CAS Number:  79-06-1
Price
156,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99.9% CAS No.: 79-06-1 Molecular Formula: CH2=CHCONH2 Formula Weight: 71.08 Flash Point: 138°(280°F) Physical Form: Crystalline Density: 1.322 Sensitivity: Air and Light Sensitive Odour: Odourless Melting Point: 82-85° Boiling Point: 125°/25mm Beilstein Reference: 605349 Merck Reference: 14,129 UN No.: 2074 MDL No.: MFCD00008032

Synonyms:  Prop-2-enamide
  • CAS Number:79-06-1
  • Formula:C₃H₅NO
  • MDL Number:MFCD00008032
  • Molecular Weight:71.08 g/mol
  • Merck Index:14,129
  • Boiling Pt:125 °C (25 mmHg)
  • Melting Pt:82…85 °C
  • Density:1.322
  • Flash Pt:138 °C (280 °F)
  • Storage Temperature:Ambient
  • EINECS:201-173-7
  • UN:2074
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications