Acrylamide ≥99.9%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99.9% CAS No.: 79-06-1 Molecular Formula: CH2=CHCONH2 Formula Weight: 71.08 Flash Point: 138°(280°F) Physical Form: Crystalline Density: 1.322 Sensitivity: Air and Light Sensitive Odour: Odourless Melting Point: 82-85° Boiling Point: 125°/25mm Beilstein Reference: 605349 Merck Reference: 14,129 UN No.: 2074 MDL No.: MFCD00008032
Synonyms: Prop-2-enamide
- CAS Number:79-06-1
- Formula:C₃H₅NO
- MDL Number:MFCD00008032
- Molecular Weight:71.08 g/mol
- Merck Index:14,129
- Boiling Pt:125 °C (25 mmHg)
- Melting Pt:82…85 °C
- Density:1.322
- Flash Pt:138 °C (280 °F)
- Storage Temperature:Ambient
- EINECS:201-173-7
- UN:2074
- ADR:6.1,III
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



