Order Entry
Export
Orders LinkContactUsLinkComponent
Urea ≥99.3%

Urea ≥99.3%

Glass bottle for solids, 500 g
Catalog # 36429.36
Urea ≥99.3%
Catalog # 36429.36
CAS Number:  57-13-6
Urea ≥99.3%
Urea ≥99.3%

Glass bottle for solids, 500 g

CAS Number:  57-13-6
Price
48,50 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99.3+% CAS No.: 57-13-6 Molecular Formula: CH4N2O Formula Weight: 60.06 Physical Form: Solid Crystalline Density: 1.335 Appearance: White Odour: Ammonia-like pH: 9,2-9,5 Melting Point: 132-135° Beilstein Reference: 635724 Merck Reference: 14,9867 MDL No.: MFCD00008022

Synonyms:  Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
  • CAS Number:57-13-6
  • Formula:NH₂CONH₂
  • MDL Number:MFCD00008022
  • Molecular Weight:60.06 g/mol
  • Merck Index:14,9867
  • Boiling Pt:197 °C (1013 hPa)
  • Melting Pt:132…135 °C
  • Density:1.335
  • Storage Temperature:Ambient
  • EINECS:200-315-5
  • REACH:01-2119463277-33
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications