Sodium borohydride ≥97%
Plastic bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Sodium borohydride ≥97%
Catalog # 35788.36
Supplier: Thermo Fisher Scientific
CAS Number: 16940-66-2
Jump to:
Product Details & Documents
Purity: 97+% CAS No.: 16940-66-2 Molecular Formula: NaBH4 Formula Weight: 37.83 Physical Form: -10+40 Mesh Granules Density: 1.074 Appearance: White Odour: Weak Melting Point: 400° dec. Solubility: Soluble in water, ammonia and amines. Insoluble in ether Merck Reference: 14,8592 UN No.: 1426 MDL No.: MFCD00003518
As a reducing agent for organic, inorganic and organometallic compounds, as a bleaching agent, in the recycling of gold and platinum metals, in organic synthesis, nonaqueous reducing agent for aldehydes, ketones, and schiff bases
Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate
- CAS Number:16940-66-2
- Formula:NaBH₄
- MDL Number:MFCD00003518
- Molecular Weight:37.83 g/mol
- Merck Index:14,8592
- Melting Pt:400 °C (dec.)
- Density:1.074
- Flash Pt:69 °C
- Storage Temperature:Ambient
- EINECS:241-004-4
- UN:1426
- ADR:4.3,I
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Cat. No.:35788.36



