Order Entry
Export
Orders LinkContactUsLinkComponent
Sodium borohydride ≥97%

Sodium borohydride ≥97%

Plastic bottle for solids, 100 g
Catalog # 35788.22
Flammable
Toxic
Corrosive
Sodium borohydride ≥97%
Catalog # 35788.22
CAS Number:  16940-66-2
Sodium borohydride ≥97%
Sodium borohydride ≥97%

Plastic bottle for solids, 100 g

CAS Number:  16940-66-2
Price
90,70 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 97+% CAS No.: 16940-66-2 Molecular Formula: NaBH4 Formula Weight: 37.83 Physical Form: -10+40 Mesh Granules Density: 1.074 Appearance: White Odour: Weak Melting Point: 400° dec. Solubility: Soluble in water, ammonia and amines. Insoluble in ether Merck Reference: 14,8592 UN No.: 1426 MDL No.: MFCD00003518

As a reducing agent for organic, inorganic and organometallic compounds, as a bleaching agent, in the recycling of gold and platinum metals, in organic synthesis, nonaqueous reducing agent for aldehydes, ketones, and schiff bases

Synonyms:  Sodium tetrahydridoborate, Sodium tetrahydroborate
  • CAS Number:16940-66-2
  • Formula:NaBH₄
  • MDL Number:MFCD00003518
  • Molecular Weight:37.83 g/mol
  • Merck Index:14,8592
  • Melting Pt:400 °C (dec.)
  • Density:1.074
  • Flash Pt:69 °C
  • Storage Temperature:Ambient
  • EINECS:241-004-4
  • UN:1426
  • ADR:4.3,I
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g

Specifications