Order Entry
Export
Orders LinkContactUsLinkComponent
Hydrochloric acid standardised solution 1.0 N

Hydrochloric acid standardised solution 1.0 N

Glass bottle, 20 L
Catalog # 35640.M4
Corrosive
Hydrochloric acid standardised solution 1.0 N
Catalog # 35640.M4
CAS Number:  7647-01-0
Hydrochloric acid standardised solution 1.0 N
Hydrochloric acid standardised solution 1.0 N

Glass bottle, 20 L

CAS Number:  7647-01-0
Price
205,00 €
Log in for availability
1 * 20 L
Product Details & Documents

CAS No.: 7647-01-0 Molecular Formula: HCl Formula Weight: 36.46 Physical Form: Liquid Appearance: Colourless to pale yellow

Synonyms:  Hydrochloric acid solution, Muriatic acid
  • CAS Number:7647-01-0
  • Formula:HCl
  • MDL Number:MFCD00011324
  • Molecular Weight:36.46 g/mol
  • Storage Temperature:Ambient
  • EINECS:231-595-7
  • UN:1789
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    20 L

Specifications