Sodium hydroxide standardised solution 0.1 N in aqueous solution
Plastic bottle, 20 L
Supplier: Thermo Fisher Scientific
Sodium hydroxide standardised solution 0.1 N in aqueous solution
Catalog # 35625.M4
Supplier: Thermo Fisher Scientific
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
CAS No.: 1310-73-2 Molecular Formula: HNaO Formula Weight: 40.00 Physical Form: Liquid MDL No.: MFCD00003548
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Density:1 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:215-185-5
- UN:1824
- ADR:8,III
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Plastic bottle
- Pk:20 L



