Order Entry
Export
Orders LinkContactUsLinkComponent
Sodium hydroxide standardised solution 0.1 N in aqueous solution

Sodium hydroxide standardised solution 0.1 N in aqueous solution

Plastic bottle, 1 L
Catalog # 35625.K2
Sodium hydroxide standardised solution 0.1 N in aqueous solution
Catalog # 35625.K2
CAS Number:  1310-73-2
Sodium hydroxide standardised solution 0.1 N in aqueous solution
Sodium hydroxide standardised solution 0.1 N in aqueous solution

Plastic bottle, 1 L

CAS Number:  1310-73-2
Price
44,80 €
Log in for availability
1 * 1 L
Product Details & Documents

CAS No.: 1310-73-2 Molecular Formula: HNaO Formula Weight: 40.00 Physical Form: Liquid MDL No.: MFCD00003548

Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Density:1 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L

Specifications