Sodium hydroxide standardised solution 0.05 N in aqueous solution
Plastic bottle, 1 L
Sodium hydroxide standardised solution 0.05 N in aqueous solution
35623.K2
: 1310-73-2
CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Physical Form: Liquid Odour: Odourless MDL No.: MFCD00003548
: Caustic soda
- :1310-73-2
- :NaOH
- :MFCD00003548
- :40 g/mol
- :1 g/cm³ (25 °C)
- :Ambient
- :215-185-5
- :1824
- :8,III
- Pack type:Plastic bottle
- Pk:1 L



