Order Entry
Export
Orders LinkContactUsLinkComponent
Silver nitrate standardised solution 0.1 N in aqueous solution

Silver nitrate standardised solution 0.1 N in aqueous solution

Glass bottle, 20 L
Catalog # 35601.M4
Corrosive
Environment
Silver nitrate standardised solution 0.1 N in aqueous solution
Catalog # 35601.M4
CAS Number:  7761-88-8
Silver nitrate standardised solution 0.1 N in aqueous solution
Silver nitrate standardised solution 0.1 N in aqueous solution

Glass bottle, 20 L

CAS Number:  7761-88-8
Price
1 640,00 €
Log in for availability
1 * 20 L
Product Details & Documents

CAS No.: 7761-88-8 Molecular Formula: AgNO3 Formula Weight: 169.87 Physical Form: Liquid Sensitivity: Light Sensitive Appearance: Colourless Odour: Odourless UN No.: 3082 MDL No.: MFCD00003414

Synonyms:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • CAS Number:7761-88-8
  • Formula:AgNO₃
  • MDL Number:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • Storage Temperature:Ambient
  • EINECS:231-853-9
  • UN:3082
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    20 L

Specifications