Order Entry
Export
Orders LinkContactUsLinkComponent
Potassium hydroxide standardised solution 0.1 N in aqueous solution

Potassium hydroxide standardised solution 0.1 N in aqueous solution

Plastic bottle, 20 L
Catalog # 35599.M4
Irritant
Potassium hydroxide standardised solution 0.1 N in aqueous solution
Catalog # 35599.M4
CAS Number:  1310-58-3
Potassium hydroxide standardised solution 0.1 N in aqueous solution
Potassium hydroxide standardised solution 0.1 N in aqueous solution

Plastic bottle, 20 L

CAS Number:  1310-58-3
Price
188,00 €
Log in for availability
1 * 20 L
Product Details & Documents

CAS No.: 1310-58-3 Molecular Formula: HKO Formula Weight: 56.11 Physical Form: Liquid Appearance: Colourless

Synonyms:  Potash caustic, Caustic potash
  • CAS Number:1310-58-3
  • Formula:KOH
  • MDL Number:MFCD00003553
  • Molecular Weight:56.11 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-181-3
  • UN:1814
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    20 L

Specifications