Orders LinkContactUsLinkComponent
Silver nitrate standardised solution 1.0 N in aqueous solution
Glass bottle, 1 L
  35597.K2
Silver nitrate standardised solution 1.0 N in aqueous solution
  35597.K2
 :  7761-88-8
Silver nitrate standardised solution 1.0 N in aqueous solution
Silver nitrate standardised solution 1.0 N in aqueous solution
Glass bottle, 1 L
 :  7761-88-8
1 260,00 €
1 * 1 L
CAS No.: 7761-88-8 Molecular Formula: AgNO3 Formula Weight: 169.87 Physical Form: Liquid Sensitivity: Light Sensitive Appearance: Colourless UN No.: 3264 MDL No.: MFCD00003414
 :  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  •  :7761-88-8
  •  :AgNO₃
  •  :MFCD00003414
  •  :169.87 g/mol
  •  :Ambient
  •  :231-853-9
  •  :3264
  •  :8,II
  • Pack type:
    Glass bottle
  • Pk:
    1 L