Potassium hydroxide standardised solution 0.5 N in aqueous solution
Plastic bottle, 1 L
Supplier: Thermo Fisher Scientific
Potassium hydroxide standardised solution 0.5 N in aqueous solution
Catalog # 35595.K2
Supplier: Thermo Fisher Scientific
CAS Number: 1310-58-3
Jump to:
Product Details & Documents
CAS No.: 1310-58-3 Molecular Formula: KOH Formula Weight: 56.11 Physical Form: Liquid Appearance: Colourless UN No.: 1814 MDL No.: MFCD00003553
Synonyms: Potash caustic, Caustic potash
- CAS Number:1310-58-3
- Formula:KOH
- MDL Number:MFCD00003553
- Molecular Weight:56.11 g/mol
- Storage Temperature:Ambient
- EINECS:215-181-3
- UN:1814
- ADR:8,III
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Plastic bottle
- Pk:1 L



