Order Entry
Export
Orders LinkContactUsLinkComponent
 

Ammonia solution 50% (v/v) in aqueous solution

4 L
Catalog # 35574.K7
Corrosive
Irritant
Ammonia solution 50% (v/v) in aqueous solution
Catalog # 35574.K7
CAS Number:  1336-21-6
Ammonia solution 50% (v/v) in aqueous solution
Ammonia solution 50% (v/v) in aqueous solution

4 L

CAS Number:  1336-21-6
Price
247,00 €
Log in for availability
1 * 4 L
Product Details & Documents

CAS No.: 1336-21-6 Molecular Formula: H5NO Formula Weight: 35.05 Physical Form: Liquid Density: 0.94 Appearance: Colourless UN No.: UN2672 MDL No.: MFCD00066650

Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Density:0.94
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specifications

  • Pk:
    4 L

Specifications