Orders LinkContactUsLinkComponent
Ammonia solution 50% (v/v) in aqueous solution
500 mL
  35574.AP
Ammonia solution 50% (v/v) in aqueous solution
  35574.AP
 :  1336-21-6
Ammonia solution 50% (v/v) in aqueous solution
Ammonia solution 50% (v/v) in aqueous solution
500 mL
 :  1336-21-6
72,70 €
1 * 500 mL
CAS No.: 1336-21-6 Molecular Formula: H5NO Formula Weight: 35.05 Physical Form: Liquid Density: 0.94 Appearance: Colourless UN No.: UN2672 MDL No.: MFCD00066650
 :  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  •  :1336-21-6
  •  :NH₃
  •  :MFCD00066650
  •  :35.05 g/mol
  •  :0.94
  •  :Ambient
  •  :215-647-6
  •  :2672
  •  :8,III
  • Pk:
    500 mL