Lead(II) oxide low silver
Glass bottle for solids, 2 kg
Supplier: Thermo Fisher Scientific
Lead(II) oxide low silver
Catalog # 33330.A3
Supplier: Thermo Fisher Scientific
CAS Number: 1317-36-8
Jump to:
Product Details & Documents
CAS No.: 1317-36-8 Molecular Formula: PbO Formula Weight: 223.19 Physical Form: Powder Density: 9.53 Odour: Odourless Merck Reference: 14,5413 UN No.: 3077 MDL No.: MFCD00011164
As storage batteries, ceramic cements and fluxes, pottery and glazes, in chromium pigments, in oil refining, in paints, and in the assay of precious metal ores
Synonyms: Lead monoxide
- CAS Number:1317-36-8
- Formula:PbO
- MDL Number:MFCD00011164
- Molecular Weight:223.2 g/mol
- Merck Index:14,5413
- Boiling Pt:1470 °C (1013 hPa)
- Melting Pt:886 °C
- Density:9.53
- Storage Temperature:Ambient
- EINECS:215-267-0
- UN:3077
- ADR:9,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:2 kg



