Order Entry
Export
Orders LinkContactUsLinkComponent
Ammonia solution 28,0 - 30,0% ACS

Ammonia solution 28,0 - 30,0% ACS

Glass bottle for solids, 250 g
Catalog # 33285.30
Corrosive
Irritant
Environment
Ammonia solution 28,0 - 30,0% ACS
Catalog # 33285.30
CAS Number:  1336-21-6
Ammonia solution 28,0 - 30,0% ACS
Ammonia solution 28,0 - 30,0% ACS

Glass bottle for solids, 250 g

CAS Number:  1336-21-6
Price
39,80 €
Log in for availability
1 * 250 g
Product Details & Documents

CAS No.: 1336-21-6 Molecular Formula: NH4OH Formula Weight: 35.05 Physical Form: Liquid Density: 0.90 Appearance: Colourless Odour: Ammonia-like pH: 12 Merck Reference: 14,494 UN No.: 2672 MDL No.: MFCD00066650

Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Merck Index:14,494
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.90
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications