Phenol 99+% stabilised ACS
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Phenol 99+% stabilised ACS
Catalog # 33213.36
Supplier: Thermo Fisher Scientific
CAS Number: 108-95-2
Jump to:
Product Details & Documents
Purity: 99+% CAS No.: 108-95-2 Molecular Formula: C6H5OH Formula Weight: 94.11 Flash Point: 79°(175°F) Physical Form: Crystalline Density: 1.071 Sensitivity: Air Sensitive Odour: Sweet Melting Point: 40-43° Boiling Point: 181° Beilstein Reference: 969616 Merck Reference: 14,7241 UN No.: 1671 MDL No.: MFCD00002143
Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene
- CAS Number:108-95-2
- Formula:C₆H₅OH
- MDL Number:MFCD00002143
- Molecular Weight:94.11 g/mol
- Merck Index:14,7241
- Boiling Pt:181 °C
- Melting Pt:40…43 °C
- Density:1.071
- Flash Pt:79 °C (175 °F)
- Storage Temperature:Ambient
- EINECS:203-632-7
- UN:1671
- ADR:6.1,II
- REACH:01-2119471329-32
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



