Oleic acid ≥99%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 112-80-1 Molecular Formula: C18H34O2 Formula Weight: 282.47 Physical Form: Liquid Density: 0.887 Refractive Index: 1.4610 Sensitivity: Air Sensitive Appearance: Colourless to pale yellow Melting Point: 13-14° Boiling Point: 286°/100mm Beilstein Reference: 1726542 Merck Reference: 14,6828 MDL No.: MFCD00064242
Synonyms: (9Z)-9-Octadecenoic acid
- CAS Number:112-80-1
- Formula:C₁₈H₃₄O₂
- MDL Number:MFCD00064242
- Molecular Weight:282.47 g/mol
- Merck Index:14,6828
- Boiling Pt:286 °C (100 mmHg)
- Melting Pt:13…14 °C
- Density:0.887
- Flash Pt:180 °C
- Storage Temperature:Ambient
- EINECS:204-007-1
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



