Order Entry
Export
Orders LinkContactUsLinkComponent
n-Undecane ≥99%

n-Undecane ≥99%

Glass bottle for solids, 250 g
Catalog # 31807.30
Health
Flammable
Irritant
n-Undecane ≥99%
Catalog # 31807.30
CAS Number:  1120-21-4
n-Undecane ≥99%
n-Undecane ≥99%

Glass bottle for solids, 250 g

CAS Number:  1120-21-4
Price
176,00 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 99% CAS No.: 1120-21-4 Molecular Formula: C11H24 Formula Weight: 156.30 Flash Point: 60°(140°F) Physical Form: Liquid Density: 0.74 Refractive Index: 1.4173 Appearance: Colourless Odour: Mild Melting Point: -26° Boiling Point: 196° UN No.: 2330 MDL No.: MFCD00008959

Synonyms:  Undecane
  • CAS Number:1120-21-4
  • Formula:H₃C(CH₂)₉CH₃
  • MDL Number:MFCD00008959
  • Molecular Weight:156.31 g/mol
  • Boiling Pt:196 °C
  • Melting Pt:–26 °C
  • Density:0.74
  • Flash Pt:60 °C (140 °F)
  • Storage Temperature:Ambient
  • EINECS:214-300-6
  • UN:2330
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications