Order Entry
Export
Orders LinkContactUsLinkComponent
n-Decane ≥99%

n-Decane ≥99%

Glass bottle for solids, 500 g
Catalog # 31209.36
Health
Flammable
n-Decane ≥99%
Catalog # 31209.36
CAS Number:  124-18-5
n-Decane ≥99%
n-Decane ≥99%

Glass bottle for solids, 500 g

CAS Number:  124-18-5
Price
193,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99+% CAS No.: 124-18-5 Molecular Formula: C10H22 Formula Weight: 142.29 Flash Point: 46°(115°F) Physical Form: Liquid Density: 0.730 Refractive Index: 1.4115 Appearance: Colourless Odour: Petrol-like Melting Point: -30° Boiling Point: 172-174° Beilstein Reference: 1696981 UN No.: 2247 MDL No.: MFCD00008954

  • CAS Number:124-18-5
  • Formula:H₃C(CH₂)₈CH₃
  • MDL Number:MFCD00008954
  • Molecular Weight:142.28 g/mol
  • Boiling Pt:172…174 °C
  • Melting Pt:–30 °C
  • Density:0.730
  • Flash Pt:46 °C (115 °F)
  • Storage Temperature:Ambient
  • EINECS:204-686-4
  • UN:2247
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications