Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,9-Decadiene ≥97%

Glass bottle for solids, 10 g
Catalog # 31207.09
Flammable
Irritant
1,9-Decadiene ≥97%
Catalog # 31207.09
CAS Number:  1647-16-1
1,9-Decadiene ≥97%
1,9-Decadiene ≥97%

Glass bottle for solids, 10 g

CAS Number:  1647-16-1
Price
147,00 €
Log in for availability
1 * 10 g
Product Details & Documents

Purity: 97% CAS No.: 1647-16-1 Molecular Formula: CH2=CH(CH2)6CH=CH2 Formula Weight: 138.25 Flash Point: 42°(107°F) Physical Form: Liquid Density: 0.75 Refractive Index: 1.4325 Boiling Point: 169° UN No.: 3295 MDL No.: MFCD00008670

Synonyms:  Deca-1,9-diene
  • CAS Number:1647-16-1
  • Formula:C₁₀H₁₈
  • MDL Number:MFCD00008670
  • Molecular Weight:138.25 g/mol
  • Boiling Pt:169 °C
  • Melting Pt:–71.8 °C
  • Density:0.75
  • Flash Pt:42 °C (107 °F)
  • Storage Temperature:Ambient
  • EINECS:216-711-6
  • UN:3295
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications