Orders LinkContactUsLinkComponent
Toluene ≥99% semiconductor grade
4x1 L, 4
  19399.M1
Toluene ≥99% semiconductor grade
  19399.M1
 :  108-88-3
Toluene ≥99% semiconductor grade
Toluene ≥99% semiconductor grade
4x1 L, 4
 :  108-88-3
182,00 €
1 * 4 items
Purity: 99% min CAS No.: 108-88-3 Molecular Formula: C6H5CH3 Formula Weight: 92.14 Flash Point: 4°(40°F) Physical Form: Liquid Density: 0.867 Refractive Index: 1.4967 Odour: Aromatic Melting Point: -93° Boiling Point: 111° Beilstein Reference: 635760 Merck Reference: 14,9529 UN No.: 1294 MDL No.: MFCD00008512
 :  Methylbenzene, Phenylmethane, FM approval
  •  :108-88-3
  •  :C₆H₅CH₃
  •  :MFCD00008512
  •  :92.14 g/mol
  •  :14,9529
  •  :111 °C
  •  :–93 °C
  •  :0.867
  •  :4 °C (40 °F)
  •  :Ambient
  •  :203-625-9
  •  :1294
  •  :3,II
  •  :05-2114615130-69
  • Pk:
    4
  • Pk Info:
    4x1 L